Learn More
Enzo Life Sciences PI3 kinase p110 alpha / p85 alpha (human), (recombinant) (20 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4360020
Encompass_Preferred
Alternative name: PI3K p110 alpha / p85 alpha. Source: Produced in insect cells. Complex of human full length p110a and p85a subunits of phosphatidylinositol 3-kinase. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2025