Learn More
Eurofins Discoverx PI3 Kinase (p110β/p85α), active, 250 µg. (14-603M)
Supplier:  Eurofins Discoverx 14603M
Encompass
Complex of N-terminal 6xHis-tagged recombinant, human full length p110α and untagged recombinant, human full length p85α. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9036