Learn More
Eurofins Discoverx PI3-Kinase (p110β/p85α) murine, 250 µg. (14-794M)
Supplier:  Eurofins Discoverx 14794M
Encompass
Complex of N-terminal 6xHis-tagged, recombinant murine p110β, full length and untagged, recombinant, murine p85α, full length. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9356