Learn More
Eurofins Discoverx PI3 Kinase (p110β/p85β) murine, 250 µg. (14-788M)
Supplier:  Eurofins Discoverx 14788M
Encompass
Complex of N-terminal 6xHis-tagged recombinant, murine full length p110β and untagged, recombinant, full length murine p85β. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9346