Learn More
Sino Biological Recombinant Human PIP5K1B Protein (Full Length, GST Tag), Active
Catalog No. 502573855
Encompass_Preferred
Recombinant full length human PIP5K1B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3855 Supplier Sino Biological Supplier No. P16102BG50
Recombinant full length human PIP5K1B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.