Learn More
Aviva Systems Biology Corp Pi4k2b Antibody - N-terminal region (ARP57203_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57203_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pi4k2b. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2610042N09Rik, 4933409G22RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-278