Learn More
Aviva Systems Biology Corp PI4KB Antibody - N-terminal region (ARP56393_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56393_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PI4KB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PI4KB. Alternate names: PI4K-BETA, PI4KIIIbeta, PI4Kbeta, PIK4CB, pi4K92, NPIK, PI4K92, PI4KBETA, PI4KIIIBETAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61840