Learn More
Cayman Chemical PI4KIII InhIbItr 3 1mg
SDP
Catalog No. 502917554
Encompass_Preferred
An inhibitor of PI4KIIIβ (IC50 = 19 nM); ~100-fold selective for PI4KIIIβ over PI4KIIα, PI4KIIβ, and PI4KIIIα, PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ, and PI3KC3, as well as 100-500-fold selective over PI3KC2α, PI3KC2β, and PI3KC2γ
Catalog No. 50-291-7554 Supplier Cayman Chemical Supplier No. 395301 MG
Add to Cart
Add to Cart
An inhibitor of PI4KIIIβ (IC50 = 19 nM); ~100-fold selective for PI4KIIIβ over PI4KIIα, PI4KIIβ, and PI4KIIIα, PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ, and PI3KC3, as well as 100-500-fold selective over PI3KC2α, PI3KC2β, and PI3KC2γ