Learn More
Cayman Chemical PI4KIIIβ Inhibitor 3, Purity: ≥98%, CAS: 1245319-54-3, 5 mg
SDP
Catalog No. 502919162
Encompass_Preferred
An inhibitor of PI4KIIIβ (IC50 = 19 nM); ~100-fold selective for PI4KIIIβ over PI4KIIα, PI4KIIβ, and PI4KIIIα, PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ, and PI3KC3, as well as 100-500-fold selective over PI3KC2α, PI3KC2β, and PI3KC2γ
Catalog No. 50-291-9162 Supplier Cayman Chemical Supplier No. 395305 MG
An inhibitor of PI4KIIIβ (IC50 = 19 nM); ~100-fold selective for PI4KIIIβ over PI4KIIα, PI4KIIβ, and PI4KIIIα, PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ, and PI3KC3, as well as 100-500-fold selective over PI3KC2α, PI3KC2β, and PI3KC2γ