Learn More
Medchemexpress LLC PI4KIIIbeta-IN-10 | 1881233-39-1 | 9290DU | 99.8% | 475.58 | 10 MG
SDP
Catalog No. 5000365008
Encompass_Preferred
PI4KIIIbeta-IN-10 is a potent and selective inhibitor of PI4KIIIβ, demonstrating an IC50 of 3.6 nM. This compound exhibits minimal off-target inhibition of related lipid kinases, showing weak inhibition of PI3KC2γ, PI3Kα, and PI4KIIIα, and less than 20% inhibition for PI4K2α, PI4K2β, and PI3Kβ at concentrations up to 20 μM. It is designed for research use only, providing a valuable tool for scientific investigations.
  • Potent PI4KIIIβ inhibitor with high selectivity
  • Minimal off-target activity against related lipid kinases
  • Available in various quantities and formulations
  • Suitable for research applications
Catalog No. 50-003-65008 Supplier Medchemexpress LLC Supplier No. 10MGHY100198
PI4KIIIbeta-IN-10 is a potent and selective inhibitor of PI4KIIIβ, demonstrating an IC50 of 3.6 nM. This compound exhibits minimal off-target inhibition of related lipid kinases, showing weak inhibition of PI3KC2γ, PI3Kα, and PI4KIIIα, and less than 20% inhibition for PI4K2α, PI4K2β, and PI3Kβ at concentrations up to 20 μM. It is designed for research use only, providing a valuable tool for scientific investigations.
  • Potent PI4KIIIβ inhibitor with high selectivity
  • Minimal off-target activity against related lipid kinases
  • Available in various quantities and formulations
  • Suitable for research applications