Learn More
Medchemexpress LLC HY-100198 1mg Medchemexpress, PI4KIIIbeta-IN-10 CAS:1881233-39-1 Purity:>98%
SDP
Catalog No. 501931744
Encompass_Preferred
Medchemexpress, HY-100198 1mg PI4KIIIbeta-IN-10 CAS:1881233-39-1 PI4KIIIbeta-IN-10 is a potent PI4KIIIβ inhibitor with an IC 50 of 3.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1744 Supplier Medchemexpress LLC Supplier No. HY1001981MG
Medchemexpress, HY-100198 1mg PI4KIIIbeta-IN-10 CAS:1881233-39-1 PI4KIIIbeta-IN-10 is a potent PI4KIIIβ inhibitor with an IC 50 of 3.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.