Learn More
Enzo Life Sciences Proteinase inhibitor 9 (human) monoclonal antibody (PI9-17) (125µg)
SDP
Catalog No. 502009913
Encompass_Preferred
Clone: PI9-17. Isotype: IgG1. Immunogen: Recombinant human full-length PI9 (proteinase inhibitor 9). Species reactivity: Human. Applications: IHC (PS), WB. Formulation: Liquid. In serum free culture supernatant containing 0.7% BSA and 0.1% sodium azide.
Catalog No. 50-200-9913 Supplier Enzo Life Sciences Supplier No. ALX804457C125
Clone: PI9-17. Isotype: IgG1. Immunogen: Recombinant human full-length PI9 (proteinase inhibitor 9). Species reactivity: Human. Applications: IHC (PS), WB. Formulation: Liquid. In serum free culture supernatant containing 0.7% BSA and 0.1% sodium azide.