Learn More
Grainger PIAB Vacuum Pad Assembly G1/8 in
Supplier:  Grainger 810UX2
Encompass_Preferred
The PIAB Vacuum Pad Assembly G1/8 in is a vacuum pad assembly with a G1/8 in threaded connection. It has a rubber vacuum pad with a diameter of 1.18 in and a height of 0.39 in. The assembly is made of anodized aluminum and has a maximum allowable vacuum of 29.5 inHg. It can handle parts weighing up to 2.2 lbs when used vertically. The pad features a grooved surface for secure gripping. This vacuum pad is suitable for material handling applications that require a small vacuum pad with a threaded connection.
Catalog No. NC2811081