Learn More
Aviva Systems Biology Corp PIAS1 Antibody - middle region (ARP39232_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39232_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIAS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PIAS1. Alternate names: DDXBP1, GBP, GU/RH-II, MGC141878, MGC141879, ZMIZ3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human
Catalog No. 50-014-56952