Learn More
Aviva Systems Biology Corp PIAS2 Antibody - C-terminal region (ARP35734_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35734_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PIAS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PIAS2. Alternate names: DIP, MIZ, MIZ1, SIZ2, ARIP3, PIASX, ZMIZ4, PIASX-BETA, PIASX-ALPHAApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-55831