Learn More
Aviva Systems Biology Corp Pias2 Antibody - N-terminal region (ARP37861_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37861_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pias2. It was validated on Western Blot. Pias2,protein inhibitor of activated STAT 2,Dib; Miz1; SIZ2; ARIP3; PIASxb; AI462206; AU018068; PIASxbeta; PIASxalpha; 6330408K17Rik; Pias2,E3 SUMO-protein ligase PIAS2,E3 SUMO-protein ligase PIAS2,DIP,PIASxalpha6,DAB2-interacting protein,Msx-interacting-zinc finger,msx-interacting zinc finger protein,protein inhibitor of activated STAT x,androgen receptor-interacting protein 3The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 6330408K17Rik, AI462206, ARIP3, AU018068, Dib, Miz1, PIASxalpha, PIASxb, PIASxbeta, SIZ2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-694