Learn More
Aviva Systems Biology Corp PIAS2 Antibody - N-terminal region (ARP53840_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53840_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIAS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PIAS2. Alternate names: MGC102682, MIZ1, PIASX-ALPHA, PIASX-BETA, SIZ2, ZMIZ4, miz, DIP, MIZ, ARIP3, PIASXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61144