Learn More
Aviva Systems Biology Corp Pias2 Antibody - C-terminal region (ARP36929_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36929_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pias2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of mouse Pias2. Alternate names: 6330408K17Rik, AI462206, ARIP3, AU018068, Dib, Miz1, PIASxalpha, PIASxb, PIASxbeta, SIZ2ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-56296