Learn More
Aviva Systems Biology Corp PIAS3 Antibody - C-terminal region (ARP32762_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32762_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PIAS3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human PIAS3. Alternate names: ZMIZ5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54747