Learn More
Aviva Systems Biology Corp PIAS3 Antibody - middle region (ARP32763_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32763_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIAS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PIAS3. Alternate names: FLJ14651, ZMIZ5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-54748