Learn More
American Radiolabeled Chemicals Inc PIB 3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 193850UC
Encompass_Preferred
6-OH-BTA-1-N-methyl-3H 50uCi in ethanol, 1 mCi/ml, 80-85 Ci/mmol, (PIB; Pittsburgh Compound B), M.W. 256.28, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-967
Add to Cart
Request a Quote