Learn More
Aviva Systems Biology Corp PIBF1 Antibody - C-terminal region (ARP52152_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52152_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIBF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PIBF1. Alternate names: C13orf24, KIAA1008, PIBF, RP11-505F3.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-651