Learn More
Cayman Chemical Pibrentasvir, Purity: ≥98%, CAS: 1353900-92-1, 10 mg
SDP
Catalog No. 502939180
Encompass_Preferred
An HCV NS5A inhibitor; inhibits HCV replication in stable replicon cell lines containing NS5A from genotypes 1-6 (EC50s = 1.4-4.3 pM); acts synergistically with glecaprevir to inhibit HCV genotype 1b-Con1 replication in replicon cells
Catalog No. 50-293-9180 Supplier Cayman Chemical Supplier No. 2754610 MG
An HCV NS5A inhibitor; inhibits HCV replication in stable replicon cell lines containing NS5A from genotypes 1-6 (EC50s = 1.4-4.3 pM); acts synergistically with glecaprevir to inhibit HCV genotype 1b-Con1 replication in replicon cells