Learn More
Cayman Chemical PIbrentasvIr 50mg
SDP
Catalog No. 502940529
Encompass_Preferred
An HCV NS5A inhibitor; inhibits HCV replication in stable replicon cell lines containing NS5A from genotypes 1-6 (EC50s = 1.4-4.3 pM); acts synergistically with glecaprevir to inhibit HCV genotype 1b-Con1 replication in replicon cells
Catalog No. 50-294-0529 Supplier Cayman Chemical Supplier No. 2754650 MG
An HCV NS5A inhibitor; inhibits HCV replication in stable replicon cell lines containing NS5A from genotypes 1-6 (EC50s = 1.4-4.3 pM); acts synergistically with glecaprevir to inhibit HCV genotype 1b-Con1 replication in replicon cells