Learn More
Selleck Chemical LLC Pibrentasvir ABT-530-5mg
SDP
Supplier:  Selleck Chemical LLC S96415MG
Encompass_Preferred
Pibrentasvir (ABT-530) is a novel and pan-genotypic hepatitis C virus (HCV) NS5A inhibitor with EC50 ranging from 1.4 pM to 5.0 pM against HCV replicons containing NS5A from genotypes 1 to 6.
Catalog No. 50-238-9611