Learn More
Waters Corp Waters Corporation Reagent, Reversed Phase, UV, Liquid, 5 pk
Catalog No. 50785910 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This PIC A Low UV Reagent analyzes ionic species via reverse-phase chromatography. It facilitates simultaneous assay of acids, bases, amphoteric, and neutral compounds, avoiding common issues like precise pH/temperature control and short column life. Designed for low UV (200 nm) analysis, it effectively eliminates background interference.
  • Separates ionic compounds via reversed-phase chromatography.
  • Enables simultaneous assay of various compound types.
  • Avoids issues with pH/temperature control and short column life.
  • Works in UV range of 200+ nm; purified and quality controlled.
  • Preformulated, buffered, and easy to use.
  • Eliminates background interference at low UV (200 nm).
  • Adjusts retention for charged and uncharged solutes.
  • 5 vials (20 mL each); room temp storage, ambient shipping.
Catalog No. 50-785-910 Supplier Waters Corp Supplier No. WAT084189
This PIC A Low UV Reagent analyzes ionic species via reverse-phase chromatography. It facilitates simultaneous assay of acids, bases, amphoteric, and neutral compounds, avoiding common issues like precise pH/temperature control and short column life. Designed for low UV (200 nm) analysis, it effectively eliminates background interference.
  • Separates ionic compounds via reversed-phase chromatography.
  • Enables simultaneous assay of various compound types.
  • Avoids issues with pH/temperature control and short column life.
  • Works in UV range of 200+ nm; purified and quality controlled.
  • Preformulated, buffered, and easy to use.
  • Eliminates background interference at low UV (200 nm).
  • Adjusts retention for charged and uncharged solutes.
  • 5 vials (20 mL each); room temp storage, ambient shipping.