Paired-Ion Chromatography (PIC™ Reagents) provide an easy solution for analyzing samples containing ionic species. These reagents allow ionic compounds to be separated by reverse-phase chromatography, saving time by allowing simultaneous assay of acids, bases, amphoteric, and neutral compounds. PIC B6 Low UV is a 5 vial/pkg of hexane sulfonic acid in a workable UV range in mobile phase 200+ nm.
- Separation mode: Reversed phase
- Detection method: UV
- Formulation: Liquid
- Storage condition: Room temperature
- System type: LC
- Shipping condition: Ambient
- Volume: 20 mL
- Type: Reagent
- Product type: Reagents
- Units per package: 5 pk
- Hazardous material: This product contains hazardous material which requires special freight handling. Additional charges may apply.