Learn More
Waters Corp Waters PIC B7 Low UV Reagent, 20 mL, 5 Vials
Catalog No. 50785913 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Paired-Ion Chromatography (PIC™ Reagents) offer a straightforward method for analyzing samples that contain ionic species. These reagents facilitate the separation of ionic compounds through reverse-phase chromatography, thereby saving time by enabling the simultaneous assay of acids, bases, amphoteric, and neutral compounds. PIC B7 Low UV is a 5 vial/package of heptane sulfonic acid, designed for use within a workable UV range of 200+ nm in the mobile phase.
  • Reversed phase separation mode
  • UV detection method
  • Liquid formulation
  • Room temperature storage condition
  • LC system type
  • Ambient shipping condition
  • Volume: 20 mL
  • 5 vials per package
  • Contains hazardous material, requiring special freight handling and potential additional charges
Catalog No. 50-785-913 Supplier Waters Corp Supplier No. WAT084282
Paired-Ion Chromatography (PIC™ Reagents) offer a straightforward method for analyzing samples that contain ionic species. These reagents facilitate the separation of ionic compounds through reverse-phase chromatography, thereby saving time by enabling the simultaneous assay of acids, bases, amphoteric, and neutral compounds. PIC B7 Low UV is a 5 vial/package of heptane sulfonic acid, designed for use within a workable UV range of 200+ nm in the mobile phase.
  • Reversed phase separation mode
  • UV detection method
  • Liquid formulation
  • Room temperature storage condition
  • LC system type
  • Ambient shipping condition
  • Volume: 20 mL
  • 5 vials per package
  • Contains hazardous material, requiring special freight handling and potential additional charges