Learn More
Waters Corp Waters Reagent, PIC B8, Low UV, 5 Vials
Catalog No. 50785914 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Paired-Ion Chromatography (PIC® Reagents) provide an easy solution for analyzing samples containing ionic species. These reagents allow ionic compounds to be separated by reverse-phase chromatography, saving time by allowing simultaneous assay of acids, bases, amphoteric and neutral compounds. PIC B8 low UV is a 5 vial/pkg of octane sulfonic acid in a workable UV range in mobile phase 200+ nm.
  • Increases retention of oppositely charged solutes.
  • Decreases retention of equally charged solutes.
  • Eliminates background interference in the low UV range.
  • Purified and quality-controlled by liquid chromatography.
  • Preformulated and buffered for appropriate pH.
  • Provides method development opportunities for ion pairs without chromophores.
  • Offers increased sensitivity potential.
Catalog No. 50-785-914 Supplier Waters Corp Supplier No. WAT084283
Paired-Ion Chromatography (PIC® Reagents) provide an easy solution for analyzing samples containing ionic species. These reagents allow ionic compounds to be separated by reverse-phase chromatography, saving time by allowing simultaneous assay of acids, bases, amphoteric and neutral compounds. PIC B8 low UV is a 5 vial/pkg of octane sulfonic acid in a workable UV range in mobile phase 200+ nm.
  • Increases retention of oppositely charged solutes.
  • Decreases retention of equally charged solutes.
  • Eliminates background interference in the low UV range.
  • Purified and quality-controlled by liquid chromatography.
  • Preformulated and buffered for appropriate pH.
  • Provides method development opportunities for ion pairs without chromophores.
  • Offers increased sensitivity potential.