Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters PIC Reagent Sample Kit, 5 Vials, Reversed Phase, Liquid, 15 mL

Catalog No. 50785935 Shop All Waters Corporation Products
Encompass_Preferred

The PIC Reagent Sample Kit is designed for paired-ion chromatography, enabling straightforward analysis of ionic species. It facilitates the separation of ionic compounds via reverse-phase chromatography, allowing simultaneous assay of acids, bases, amphoteric, and neutral compounds, thereby saving time.

  • Designed for analyzing samples containing ionic species.
  • Facilitates separation of ionic compounds using reverse-phase chromatography.
  • Enables simultaneous assay of acids, bases, amphoteric, and neutral compounds.
  • Separation mode is reversed phase.
  • Detection method is UV.
  • Formulation is liquid.
  • Storage condition is room temperature.
  • System type is LC.
  • Shipping condition is ambient.
  • Each vial has a volume of 15 mL.
  • Type is reagent.
  • Product type is application kits.

Catalog No. 50-785-935 Supplier Waters Corp Supplier No. WAT085144
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The PIC Reagent Sample Kit is designed for paired-ion chromatography, enabling straightforward analysis of ionic species. It facilitates the separation of ionic compounds via reverse-phase chromatography, allowing simultaneous assay of acids, bases, amphoteric, and neutral compounds, thereby saving time.

  • Designed for analyzing samples containing ionic species.
  • Facilitates separation of ionic compounds using reverse-phase chromatography.
  • Enables simultaneous assay of acids, bases, amphoteric, and neutral compounds.
  • Separation mode is reversed phase.
  • Detection method is UV.
  • Formulation is liquid.
  • Storage condition is room temperature.
  • System type is LC.
  • Shipping condition is ambient.
  • Each vial has a volume of 15 mL.
  • Type is reagent.
  • Product type is application kits.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.