Learn More
Aviva Systems Biology Corp PICALM Antibody - C-terminal region (ARP63958_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63958_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PICALM. It was validated on Western Blot. . Alternate names: CALM, CLTH, LAPApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-131