Learn More
FUJIFILM Biosciences Picarbutrazox Metabolite B Sta
Catalog No. 5000450410
Encompass_Preferred
Reagent for Pesticide Residue Analysis Used in pesticide residue analysis supplied as a white powder with minimum 95% assay via HPLC/qNMR soluble in acetone
Catalog No. 50-004-50410 Supplier FUJIFILM Biosciences Supplier No. 16328401
Reagent for Pesticide Residue Analysis Used in pesticide residue analysis supplied as a white powder with minimum 95% assay via HPLC/qNMR soluble in acetone