Learn More
Cayman Chemical Picaridin-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2928067-33-6, 1 mg
SDP
Catalog No. 502949061
Encompass_Preferred
An internal standard for the quantification of picaridin by GC- or LC-MS
Catalog No. 50-294-9061 Supplier Cayman Chemical Supplier No. 344241 MG
An internal standard for the quantification of picaridin by GC- or LC-MS