Learn More
Huber USA PICCOLO 280 OLE
Catalog No. NC1947819
Encompass_Preferred
With new controller OLE - Refrigerated Heating Circulator, temperature range: 4 ... 70 °C, Cooling/ heating capacity: at 40°C: 0,4 kWat 10°C: 0,1 kW, prsurre pump max: 1,7 l/min / 0.8 bar, Pump connection: Connector with hose barb, Power supply: 100-240V 1~/2~ 50/60Hz.
Catalog No. NC1947819 Supplier Huber USA Supplier No. 3044.0002.98
Add to Cart
Add to Cart
Request a Quote
With new controller OLE - Refrigerated Heating Circulator, temperature range: 4 ... 70 °C, Cooling/ heating capacity: at 40°C: 0,4 kWat 10°C: 0,1 kW, prsurre pump max: 1,7 l/min / 0.8 bar, Pump connection: Connector with hose barb, Power supply: 100-240V 1~/2~ 50/60Hz.