Learn More
Enzo Life Sciences Piceatannol (10mg). CAS: 10083-24-6
SDP
Catalog No. 502008421
Encompass_Preferred
Originally isolated from Euphorbia lagascae. Selective protein tyrosine kinase Syk inhibitor (IC50~10 µM). In RBL-2H3 cells the selective inhibition of Syk by piceatannol results in inhibition of FcεR1-mediated signaling. Activator of human deacetylase SIRT1. Alternative name: 3,4,3',5'-Tetrahydroxy-trans-stilbene. Formula: C14H12O4. MW: 244.2. Purity: ≥98% (HPLC (UV)). Solubility: Soluble in DMSO (>50mg/ml), methanol (>50mg/ml), dioxan (>50mg/ml), acetonitrile (>50mg/ml); slightly soluble in water (1mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8421 Supplier Enzo Life Sciences Supplier No. ALX270202M010
Originally isolated from Euphorbia lagascae. Selective protein tyrosine kinase Syk inhibitor (IC50~10 µM). In RBL-2H3 cells the selective inhibition of Syk by piceatannol results in inhibition of FcεR1-mediated signaling. Activator of human deacetylase SIRT1. Alternative name: 3,4,3',5'-Tetrahydroxy-trans-stilbene. Formula: C14H12O4. MW: 244.2. Purity: ≥98% (HPLC (UV)). Solubility: Soluble in DMSO (>50mg/ml), methanol (>50mg/ml), dioxan (>50mg/ml), acetonitrile (>50mg/ml); slightly soluble in water (1mg/ml). Long Term Storage: +4°C.