Learn More
Apexbio Technology LLC Piceatannol 10083-24-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063299
Encompass_Preferred
Piceatannol (CAS 10083-24-6) is a naturally occurring small molecule exhibiting anti-inflammatory immunomodulatory and antiproliferative activities It functions primarily by inhibiting protein tyrosine kinases including p56lck and syk kinases and suppressing tumor necrosis factor (TNF)-induced nuclear factor-kappa B (NF- B) activation and downstream gene expression Biologically piceatannol arises as a metabolite of resveratrol through oxidation by cytochrome P450 1B1 This compound is employed in biomedical research to explore kinase signaling pathways inflammatory response regulation and potential therapeutic interventions
Catalog No. 50-000-63299 Supplier Apexbio Technology LLC Supplier No. N20315.1
Piceatannol (CAS 10083-24-6) is a naturally occurring small molecule exhibiting anti-inflammatory immunomodulatory and antiproliferative activities It functions primarily by inhibiting protein tyrosine kinases including p56lck and syk kinases and suppressing tumor necrosis factor (TNF)-induced nuclear factor-kappa B (NF- B) activation and downstream gene expression Biologically piceatannol arises as a metabolite of resveratrol through oxidation by cytochrome P450 1B1 This compound is employed in biomedical research to explore kinase signaling pathways inflammatory response regulation and potential therapeutic interventions