Learn More
Apexbio Technology LLC Piceatannol, 20mg. Cas: 10083-24-6 MFCD: MFCD00221715. p56lck/Syk inhibitor
SDP
Catalog No. 501905649
Encompass_Preferred
Anti-inflammatory, immunomodulatory and antiproliferative agent. Inhibits p56lck and syk protein tyrosine kinases and inhibits TNF-induced NF-B activation and gene expression. Synthesis results from conversion of resveratrol by cytochrome P450 1B1. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5649 Supplier Apexbio Technology LLC Supplier No. N203120
Anti-inflammatory, immunomodulatory and antiproliferative agent. Inhibits p56lck and syk protein tyrosine kinases and inhibits TNF-induced NF-B activation and gene expression. Synthesis results from conversion of resveratrol by cytochrome P450 1B1. Other sizes are available. Please inqury us for quote.