Learn More
TARGETMOL CHEMICALS INC PICEATANNOL 3-O-GLUCOSID 5MG
SDP
Catalog No. 502580017
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Piceatannol 3'-O-glucoside (Piceatannol 3-glycoside) increases pulmonary blood oxygen tension and decreases pulmonary interstitial edema so it can protect lung injury. purity: 99%
Catalog No. 50-258-0017 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S04015MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Piceatannol 3'-O-glucoside (Piceatannol 3-glycoside) increases pulmonary blood oxygen tension and decreases pulmonary interstitial edema so it can protect lung injury. purity: 99%