Learn More
Medchemexpress LLC Picfeltarraenin IA | 97230-47-2 | 762.92 | 5 MG
SDP
Catalog No. 5000415123
Encompass_Preferred
Picfeltarraenin IA is a triterpenoid derived from Picriafel-terrae Lour (P.fel-terrae). It acts as an acetylcholinesterase (AChE) inhibitor and is suitable for research on herpes infections, cancer, and inflammation.
  • Acts as an acetylcholinesterase inhibitor
  • Derived from Picriafel-terrae Lour
  • Used in research for herpes infections
  • Used in research for cancer
  • Used in research for inflammation
Catalog No. 50-004-15123 Supplier Medchemexpress LLC Supplier No. HYN14745MG
Picfeltarraenin IA is a triterpenoid derived from Picriafel-terrae Lour (P.fel-terrae). It acts as an acetylcholinesterase (AChE) inhibitor and is suitable for research on herpes infections, cancer, and inflammation.
  • Acts as an acetylcholinesterase inhibitor
  • Derived from Picriafel-terrae Lour
  • Used in research for herpes infections
  • Used in research for cancer
  • Used in research for inflammation