Learn More
Teknova Pichia Drop Out Adenine Selective Agar Plates 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Supplier:  Teknova P11104PK
Encompass
Pichia Selective Agar,Carbon: 2.0% Cerelose Anhydrous Dextrose (Glucose),Nitrogen: 1.0% Ammonium Sulfate,Minus: Adenine
Catalog No. 50-238-7744