Learn More
Cygnus Technologies PICHIA PASTORIS HCP ELISA KIT
Supplier:  Cygnus Technologies F140
Encompass
This kit is intended for use in determining the presence of host cell protein contamination in products manufactured by recombinant expression in Pichia pastoris. This kit has an LOD of <0.3ng/mL.
Catalog No. NC9495630