Learn More
American Research Products Inc Pichia Yeast host cell protein
Catalog No. 5000326407
Encompass
Pichia Yeast host cell protein (HCP) Residue ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-800 ng/mL Sensitivity 1 56 ng/mL Sample cell culture supernates products produced during process of protein purification and final products Method Sandwich
Catalog No. 50-003-26407 Supplier American Research Products Inc Supplier No. CSBEQ33265PY
Pichia Yeast host cell protein (HCP) Residue ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-800 ng/mL Sensitivity 1 56 ng/mL Sample cell culture supernates products produced during process of protein purification and final products Method Sandwich