Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PichiaPink™ Secreted Protein Kit

Catalog No. A11151
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11151 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A11151 Supplier Invitrogen™ Supplier No. A11151
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PichiaPink™ Secreted Protein Kit

The PichiaPink™ Secreted Protein Expression Kit is a new recombinant protein expression system based on the yeast Pichia pastoris. The PichiaPink™ Secreted Protein Expression Kit gives you convenient and cost-efficient bioproduction from shake flask to 1000-liter production of your recombinant protein. The PichiaPink™ Secreted Protein Expression Kit comes with our new Pichia strains, the pPINK-HC and pPINK-LC vectors, and secretion signal sequences for production of your protein.
  •   Get started with secreted protein production in Pichia pastoris.
  •   Screen your Pichia transformants rapidly by auxotrophy and color selection.
  •   Reduce protein degradation with protease-deficient Pichia pastoris strains.

    The PichiaPink™ Secreted Protein Expression Kit comes with:
  •   PichiaPink™ Secreted Protein Vector Kit (Cat. No. A11153)
  •   PichiaPink™ Expression Strain Set (Cat. No. A11154)
  •   PichiaPink™ Media Kit (Cat. No. A11156)

    A New Way to Screen Pichia Transformants
    The PichiaPink™ Secreted Protein Expression Kit uses adenine auxotrophy to select for Pichia transformants. The PichiaPink™ strains have the ade2 genotype and require complementation with the ADE2 gene to grow without adenine. Use of an auxotrophic marker also makes it easier to maintain transformants. No antibiotics are required to maintain your transformants.

    In addition, you can screen for Pichia transformants by color. The untransformed PichiaPink™ strains appear as pink colonies while transformed PichiaPink™ strains appear as white colonies (Fig 1).

    Secrete Your Proteins with the pPINKα-HC Vector

    The PichiaPink™ Secreted Protein Expression Kit comes with the pPINKα-HC vector. The pPINKα-HC vector has the α-mating factor signal sequence built-in and allows your protein to be secreted into the media. Secreting your protein into the media makes harvesting and purifying your protein easier. The pPINKα-HC vector is built around the ADE2 gene for complementing the ade2-deficient Pichia strains. This vector uses the methanol-induced AOX1 promoter to express your protein. Further, the pPINKα-HC vector is designed to be integrated at high-copy-number in your Pichia transformants.

    The PichiaPink™ Secreted Protein Vector Kit (Cat. No. A11153) can be ordered separately to refill the PichiaPink™ Secreted Protein Expression Kit.

    Keep Your Proteins Intact
    The PichiaPink™ Secreted Protein Expression Kit comes with the PichiaPink™ Expression Strain Set. This set contains the 4 Pichia pastoris strains for use with the PichiaPink™ Yeast Expression System. All 4 strains have the ade2 genotype. In addition, 3 strains include knockouts in 2 protease genes. Protease-deficient strains reduce the need for protease inhibitors when growing your cells and improve the yield of your protein. Test a single protease knockout or the double protease knockout to find the one that works best for you.

    You can order the PichiaPink™ Expression Strain Set (Cat. No. A11154) separately to refill the PichiaPink™ Secreted Protein Expression Kit .

    Media Pouches to Get You Started

    The PichiaPink™ Media Kit is a set of convenient prepackaged media pouches for use with the PichiaPink™ Secreted Protein Expression Kit. The PichiaPink™ Media Kit contains media needed to grow PichiaPink™ strains from frozen stocks to cultures ready for transformation and selection. You can order the PichiaPink™ Media Kit (Cat. No. A11156) separately to refill the PichiaPink™ Secreted Protein Expression Kit.

    Why Choose the PichiaPink™ Yeast Expression System?
    The PichiaPink™ Yeast Expression System is based on the yeast Pichia pastoris. Advantages of Pichia pastoris include rapid growth, well-defined genetic background, simple media formulation, and easy handling. For over 30 years, Pichia pastoris has been used by labs around the world for producing hundreds of different proteins from many species including human (Ref 1, 2). The PichiaPink™ Yeast Expression System allows convenient and cost-effective protein production from small to large scales.

    For information on obtaining a commercial-use license for the PichiaPink™Yeast Expression System, please inquire at outlicensing@lifetech.com.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

    Related Links
  • Learn more about the PichiaPink™ Yeast Expression System.
  • Learn more about our other protein expression systems.

    References

    1. Cereghino JL, Cregg JM. Heterologous protein expression in the methylotrophic yeast Pichia pastoris. FEMS Microbiol Rev. 2000 Jan;24(1):45-66. [PubMed]
    2. Cereghino GP, Cereghino JL, Ilgen C, Cregg JM. Production of recombinant proteins in fermenter cultures of the yeast Pichia pastoris. Curr Opin Biotechnol. 2002 Aug;13(4):329-32. [PubMed]
  • TRUSTED_SUSTAINABILITY

    Specifications

    Type Pichia Protein Expression System
    Quantity 1 kit
    Content And Storage The PichiaPink™ Secreted Protein Expression kit consists of:
    • The PichiaPink™ Secreted Protein Vector kit containing the high-copy number plasmid with the S. cerevisiae Alpha mating factor presequence to yield secreted protein expression.
    • The PichiaPink™ Expression Strain set containing 4 glycerol strains
    • The PichiaPink™ Media Kit containing 4 basic media.
    Compatible Cells Pichia pastoris strains
    Shipping Condition Room Temperature
    Expression Mechanism Cell-Based Expression
    Expression System Yeast
    Product Line PichiaPink
    Species P. pastoris
    Expression Stability Stable
    Final Product Type Liquid Medium, Agar Plates
    Show More Show Less
    When selecting for blasticidin-resistant transformants in the X-33 strain using pPIC6/pPIC6α vectors, why do I get large and small colonies on YPD plates containing 300 µg/ml blasticidin?

    Generally, large colonies represent transformants containing pPIC6/pPIC6α integrants, while small colonies represent transformants containing pPIC6/pPIC6α non-integrants. These non-integrants have transduced the pPIC6/pPIC6α plasmid, and therefore, exhibit a low level of blasticidin resistance in the initial selection process. Upon subsequent screening, these non-integrant transformants do not retain blasticidin resistance.

    When choosing a blasticidin-resistant transformant for your expression studies, we recommend that you pick blasticidin-resistant colonies from the initial transformation plate and streak them on a second YPD plate containing the appropriate concentration of blasticidin. Select transformants that remain blasticidin-resistant for further studies.

    My transformation is not working. Do you have any suggestions?

    Here are some suggestinos:

    - Make sure that you have harvested cells during log-phase growth (OD <1.0 generally).
    - If electroporation is being used, see the electroporator manual for suggested conditions. Vary electroporation parameters if necessary.
    - Use more DNA.
    - Use freshly made competent cells.
    - If the LiCl transformation method is being used, try boiling the carrier DNA.

    My spheroplasting of Pichia worked twice, but hasn't worked since. The OD of the culture simply does not drop.

    Here are some things to consider:

    - If the OD of cells that are used is too high, they will not spheroplast. Do not overgrow cells.
    - Do not use old cells and make sure that they are in log phase of growth.
    - Make sure to mix zymolyase well before using. Zymolyase is more of a suspension than a solution.
    - Make the PEG solution fresh each time and check the pH.

    Is there a recommended protocol for fermentation using constitutive expression vectors such as pGAPZ?

    Use the following high cell density protocol for pGAP clones. Feed carbon until the desired density is reached (300 to 400 g/L wet cell weight (WCW)). If the protein is well-behaved in the fermenter, increase to 300-400 g/L WCW as with methanol inducible clones. These densities can be reached in less than 48 hours of fermentation. We have fermented constitutive expressers on glycerol using these protocols with good results. Some modifications to the Fermentation Basal Salts Medium that you might want to make are:

    1) Substitute 2% dextrose for the 4% glycerol in the batch medium.
    2) Substitute 40% dextrose for the 50% glycerol in the fed-batch medium.
    3) Feed the 40% dextrose at 12 mL/L/hr (Jim Cregg has published data on expression using several carbon sources as substrates; dextrose gave the highest levels of expression).
    4) Yeast extract and peptone may be added to the medium for protein stability.

    One warning: If you are working with His- strains, they remain His- after transformation with pGAPZ. Fermentation in minimal medium will require addition of histidine to the fermenter.

    Can the methanol and ammonium hydroxide solutions used to prepare Pichia fermentation medium be autoclaved?

    No, you cannot autoclave methanol. There are two approaches to this, depending a bit on the size of the bioreactor and the volumes involved. You can either dilute to working concentration and filter-sterilize with a filter suitable for alcohols, or you can just assume that methanol is sterile (it should be) and dilute into sterile water. For the ammonium hydroxide solution, you should also not autoclave it. You can assume the 30% stock solution is sterile (nothing should live in this solution) and dilute into sterile water to the working concentration.

    Can antibiotics be used during Pichia fermentation?

    The use of antibiotics is not recommended, because most antibiotics become inactivated at the low pH of the medium during Pichia fermentation. In other words, addition of antibiotics such as ampicillin or kanamycin won't hurt the fermentation process, but because of the low pH the antibiotics become inactivated or may even precipitate out. For best results, use good sterile techniques.

    Do I need to add sulfuric acid to the fermentation PTM trace salts?

    You don't have to add sulfuric acid to your PTM1 salts or fermentation medium. It would serve no purpose, other than maybe help dissolve the salts.

    Can I use YPD instead of BMGY-type media for Pichia fermentation?

    Yes. The cells will do fine in YPD, but there are two drawbacks: The foaming that occurs in the richer YPD is very difficult to control, and the richer medium makes it difficult to purify secreted proteins from the medium. The BMGY formulation remedies both of these problems.

    What is the advantage of mixed feed in Pichia fermentation?

    The use of mixed feeds is mainly due for "turning down" the level of expression for proteins that are troublesome for Pichia. We have generally used mixed feeds for MutS clones. The idea is to keep the culture in a state of more active growth, and thus "happier" to express proteins.

    What can be used as an acid to adjust the pH of Pichia fermentation media? Do I even need to adjust the pH?

    You need not add any acid to Pichia fermentation media. A healthy culture always acidifies the medium. If the pH of the culture is increasing, it is a sign of carbon source depletion or ill health of the culture.

    What pattern of oxygen uptake should I expect to observe during a Pichia fermentation run?

    It depends whether the clone is Mut+ or a MutS.

    For a Mut+ clone, you should expect that initially (in the first 2-4 hours of induction), the oxygen uptake rate of the culture would be lower than that at the end of the glycerol batch phase. After the culture becomes adapted to methanol, the oxygen uptake rate will significantly increase, if the culture is healthy (i.e., not poisoned by too much methanol). One should run methanol spike tests during fermentation of Mut+ clones.

    For a MutS clone, one can expect that the oxygen uptake rate will be lower than that at the end of the glycerol batch phase throughout most of the fermentation. One has to be very careful not to poison MutS clones.

    Do you have any protocols for Pichia fermentation?

    We do not offer any protocols for Pichia fermentation. Please refer to the document titled “Pichia Fermentation Guidelines” on our website.

    Is there an electroporation protocol for Pichia cells that doesn't require starting with 500 mL of cells?

    The following protocol has been used numerous times for Pichia pastoris. It uses a 250 mL culture that is eventually scaled down to 1 mL aliquots of each strain.

    - Inoculate 10 mL YPD media with Pichia strain and grow O/N, shaking at 30 degrees C.
    - In the morning, check the OD600. To get them in log phase by the afternoon, dilute cells to hit an OD600 of approximately 3.0 at 4 or 5 pm.
    - When the OD600 reaches approximately 3.0, inoculate 250 mL of YPD with 250 µL of culture. The objective is to have healthy, log-phase cells in the morning at an OD600 of around 1.0.
    - If the OD600 is ~1.0, spin the cells in a 1 L bottle at 3K rpm for 10 minutes.
    - Gently resuspend in 250 mL cold dH20.
    - Transfer to a 500 mL centrifuge bottle and spin at 3K for 10 min. Repeat.
    - Resuspend in 20 mL cold 1 M sorbitol and transfer to a 50 mL conical tube.
    - Spin at 3K rpm for 10 min.
    - Resuspend in 1 mL 1M sorbitol, and keep on ice.
    - Use 80 µL of host strain for each electroporation.

    What is the purpose of including sorbitol in the YPD plates used for plating Pichia cells after electroporation?

    Inclusion of 1 M sorbitol in YPD plates stabilizes electroporated cells, as they appear to be somewhat osmotically sensitive.

    Is it critical that one uses PEG 4000 for yeast transformations?

    PEG 4000 seems to work best for yeast transformations, although PEG 3350 has been used in-house with success.

    Which method do you recommend using for transformation of Pichia?

    We recommend electroporation for transformation of Pichia. Electroporation yields 10e3 to 10e4 transformants per µg of linearized DNA and does not destroy the cell wall of Pichia. If you do not have access to an electroporation device, you may use the Spheroplast Kit for Yeast(Cat. No. K172001), PEG 1000 protocol (page 78 of the manual), LiCl protocol (page 80 of the manual), or the Pichia EasyComp Transformation Kit (Cat. No. K173001). We do not recommend spheroplasting for transformation of Pichia with plasmids containing an antibiotic resistance marker. Damage to the cell wall leads to increased sensitivity to the antibiotic, causing putative transformants to die before they express the antibiotic resistance gene. In contrast, spheroplasting can be used for transformation of PichiaPink vectors because these vectors are selected using auxotrophic markers.

    What are the different methods available for transformation of Pichia, and how do they compare?

    Here are the different methods available for Pichia transformation:

    Pichia EasyComp Transformation Kit: easy-to-use, ready-made reagents
    This method produces chemically competent Pichia cells and provides a rapid and convenient alternative to electroporation. Transformation efficiency is low (transformation of 50 µl of competent cells with 3 µg of linearized plasmid DNA yields about 50 colonies), and hence it is very difficult to isolate multi-copy integrants. Higher transformation efficiencies are often obtained with frozen versus freshly prepared cells.

    PEG 1000 transformation: easy, do-it-yourself protocol
    It is critical to add DNA to frozen cell samples, as cell competence decreases very rapidly after the cells thaw-even when held on ice. To perform multiple transformations, it is recommended to process them in groups of six at a time. The PEG method is usually better than LiCl, but not as good as spheroplasting or electroporation for transformation. However, it is convenient for people who do not have an electroporation device. The transformation efficiency is 10e2 to 10e3 transformants per mg of DNA.

    Lithium chloride transformation: easy, do-it-yourself protocol
    This method is an alternative to transformation by electroporation. Competent cells must be made fresh. Transformation efficiency is 10e2 to 10e3 transformants per µg linearized DNA. Note: Lithium acetate does not work with Pichia pastoris. Use only lithium chloride.

    Electroporation: easy and high efficiency, do-it-yourself protocol; does not destroy the cell wall
    Competent cells must be made fresh. Transformation efficiency is 10e3 to 10e4 transformants per µg of linearized DNA.

    Spheroplast Kit for Yeast (K172001): cell wall digested to allow DNA to enter the cell; the procedure involves treating cells with zymolyase to create spheroplasts.
    You must determine the optimal time to treat with zymolyase by taking OD600 readings at increasing time points. Longer incubations with zymolyase result in reduced transformation efficiency. Spheroplasts are combined with DNA and then plated. Transformation efficiency is 10e3 to 10e4 transformants per µg of linearized DNA. Note: Spheroplasting is not recommended for Pichia vectors with an antibiotic resistance marker. Damage to the cell wall leads to increased sensitivity to the antibiotic, causing putative transformants to die before they express the antibiotic resistance gene. In contrast, spheroplasting can be used for transformation of PichiaPink vectors, because these vectors are selected using auxotrophic markers.

    What is the mating genotype of your Pichia strains?

    All of our Pichia strains are homothallic strains. This means that they actually switch mating type with each generation. In Saccharomyces strains, this would lead to the culture rapidly becoming entirely diploid. In contrast, Pichia pastoris strains mate inefficiently to form diploids. Therefore, at any given time, the cells in the population are both “a” and “alpha” mating types.

    Does Pichia pastoris secrete proteins that can be toxic to itself or to other cells?

    Certain yeast strains secrete a protein toxin, which inhibits the growth of sensitive pathogens and yeasts. Studies have shown that production of the toxin is dependent on the presence of linear, double-stranded DNA plasmids in the killer yeasts. In the yeast Pichia pastoris, two linear double-stranded DNA plasmids have been identified. In the publication listed below, the search for toxin-producing capability in P. pastoris was conducted and no killer activity could be detected when 14 different indicator strains were tested. Reference: Banerjee and Verma (2000) Search for a Novel Killer Toxin in Yeast Pichia pastoris. Plasmid 43:181-183.

    Will Pichia pastoris vectors (e.g., pPICZ, pPIC6, pPIC9K, pPIC3.5K, pAO815) work in Pichia methanolica? Is the TEF1 promoter functional in Pichia methanolica?

    No, Pichia pastoris vectors will not work in Pichia methanolica; both Pichia pastoris and Pichia methanolica vectors have promoters derived from alcohol oxidase but they are not homologous, so the Pichia pastoris vectors will not be able to integrate or replicate in Pichia methanolica. The TEF1 promoter is probably functional in Pichia methanolica.

    When doing Pichia expression from a plasmid containing the Zeocin antibiotic resistance gene, is it necessary to have Zeocin antibiotic in the expression medium?

    There is no need for maintaining Zeocin antibiotic selection in the Pichia expression medium, since Pichia pastoris transformants are stable integrants with the gene of interest integrated into the genome.

    Will a recombinant protein be glycosylated as it goes through the secretory pathway in a yeast cell?

    A secreted protein will be exposed to the glycosylation machinery and might be glycosylated if the protein contains the standard N-linked or O-linked glycosylation amino acid consensus sequence.

    Can I express complex proteins or one with a quaternary structure in Pichia pastoris?

    Pichia is capable of correctly assembling proteins with a quaternary structure. One of the earliest proteins to be expressed in Pichia was the Hepatitis B Surface antigen which was assembled in its natural form, the 22 nm particle. (Reference: Cregg JM et al. (1987) High-level expression and efficient assembly of hepatitis B surface antigen in the methylotrophic yeast P. Pastoris. Nat Biotechnol 5:479-485.)  In consideration of the particle assembly problem, Cregg postulated that one or more post-translational events important in the formation of particles may be slow relative to the synthesis of HBsAg protein. Therefore, he used MutS since it has a slower growth rate.

    What do I do if I see a volume loss during a pilot expression of my Pichia culture?

    You can supplement with 10% culture volume of a 5% methanol (in water) solution to regenerate the 0.5% methanol concentration each day.

    If there are no Zeocin antibiotic-containing YPD plates readily available, would it be possible to spread Zeocin antibiotic on top of YPD plates and still retain efficient selection of yeast?

    Zeocin antibiotic can be spread on top of YPD plates for selection of yeast if necessary. There is a report that this works well when done with 10-15 3 mm glass beads. However, it is recommended that some optimization be performed, since top-spreading may dilute the antibiotic's effectiveness.

    Will the alpha factor secretion signal work in other yeast?

    The alpha secretion signal is from S. cerevisiae and is a general yeast secretion signal that has been used in many species including P. pastoris, K. lactis, etc.

    How is the alpha factor secretion signal sequence processed?

    The alpha “signal sequence” (which really contains both the alpha signal sequence and pro-hormone leader sequences) is cleaved 4 times by 3 different enzymes in the Pichia cell. First, near the N-terminus by signal peptidase; second, by Kex2p after the dibasic (Lys-Arg) signal slightly upstream of the multiple cloning site, and then twice by Ste13p to remove the 2 Glu-Ala repeats.

    Will a mammalian secretion signal work in Pichia?

    Although the efficiency may differ from one signal to the next, in general mammalian secretion signals are functional in yeast.

    What is the codon usage for Pichia?

    It is doubtful as to whether codon usage plays as great a role in general, as is commonly believed. Translation initiation is probably more of a rate-limiting step than elongation.
    Use the following codon usage list to design your gene in the order of preference:

    Glycine: GGT or GGA
    Glutamic acid: GAG or GAA
    Aspartic acid: GAC or GAT
    Valine: GTT or GTC
    Alanine: GCT or GCC
    Arginine: AGA or CGT
    Serine: TCT or TCC
    Lysine: AAG
    Asparagine: AAC
    Methionine: ATG
    Isoleucine: ATT or ATC
    Threonine: ACT or ACC
    Tryptophan: TGG
    Cysteine: TGT
    Tyrosine: TAC
    Leucine: TTG or CTG
    Phenylalanine: TTC
    Glutamine: CAA or CAG
    Histidine: CAC or CAT
    Proline: CCA or CCT

    How can I convert OD units to approximate Pichia cells/mL (or density)?

    An OD600 of 1 is equivalent to 5 x 10e7 Pichia cells/mL. After overnight (O/N) growth from a colony pick, a Pichia culture generally reaches OD 1.3-1.5 (in 2-5 mL).

    What is the doubling time of Pichia? How long should I wait to see colonies on agar?

    Doubling time is 2-3.5 hrs: Pichia has a doubling time of about 2 hrs of glycerol. The yeast grow slowly at 30 degrees C and it takes at least 3 days for colonies. In practice, it takes anywhere from 3 to 7 days to get nice-sized colonies.

    What is the size of the Pichia genomic DNA?

    The Pichia genome is similar to that of other yeast, approximately 1.5 x 107 bp (similar to S. cerevisiae) and contains 4 chromosomes (similar to S. pombe). Reference: Ohi H, Okazaki N, Uno S, Miura M, Hiramatsu R (1998) Chromosomal DNA patterns and gene stability of Pichia pastoris. Yeast 14(10):895-903.

    We have clearly resolved four chromosomal bands from four Pichia pastoris (Komagataella pastoris) strains by using contour-clamped homogeneous electric field gel electrophoresis. The size of the P. pastoris chromosomal bands ranged from 1.7 Mb to 3.5 Mb, and total genome size was estimated to be 9.5 Mb to 9.8 Mb; however, chromosome-length polymorphisms existed among four strains.

    What selection mechanism does the PichiaPink Yeast Expression System use?

    The PichiaPink system relies on selection of transformants using ADE2 complementation (i.e., by complementation of adenine auxotrophy) rather than antibiotic selection. The ADE2 gene encodes phosphoribosylaminoimidazole carboxylase, which catalyzes the sixth step in the de novo biosynthesis of purine nucleotides. Mutations in ADE2 lead to the accumulation of purine precursors in the vacuole, which causes the colony to be red in color. In addition, ade2 mutants are adenine auxotrophs that are unable to grow on medium lacking adenine and have a slow growth phenotype on rich medium.

    The strains in the PichiaPink system are ade2 auxotrophs due to the full deletion of the ADE2 gene and part of its promoter. The PichiaPink expression vectors contain the ADE2 gene (under its own promoter) as the selection marker, with the high-copy vectors (pPink-HC and pPinkalpha-HC) containing a truncated ADE2 promoter compared to the full-length ADE2 promoter in the low-copy vector (pPink-LC). Transformation of the PichiaPink strains with the expression plasmids enable the strain to grow on medium lacking adenine (Ade dropout medium or minimal medium). Regardless of the host PichiaPink strain, both white and slightly pink colonies are obtained on the selection plates upon transformation with the high-copy PichiaPink vectors. The color of the colonies indirectly indicates the relative expression levels of the protein of interest as the color of the colony depends on the copy number of the plasmid, which in turn is determined by the promoter strengths of the markers. The pink colonies express very little ADE2 gene product, while the white colonies express higher amounts of the ADE2 gene product, suggesting that those colonies have more copies of the integrated construct. Strains transformed with the low-copy plasmid, pPink-LC, grow faster on medium lacking adenine, generating white colonies due to the stronger promoter on this vector. Since the promoter is stronger, less ADE2 expression is required to allow the strains to grow on medium lacking adenine. As a result, fewer copies of the ADE2 gene/expression construct are required in the strain.

    What are the advantages of the PichiaPink Yeast Expression System over the EasySelect Yeast Expression system?

    PichiaPink Yeast Expression System offers significant advantages compared to the original EasySelect Pichia system. Please see the advantages below:

    - Both high and low copy enables optimization of toxic protein expression
    - 8 secretion signal leader sequences
    - 4 strains
    - 3 protease-deficienct host strains
    - Relies on adenine selection instead of an antibiotic resistance marker

    How do I store yeast long term? Can I freeze them at -80 degrees C like bacteria? What is the shelf life of frozen stocks?

    We recommend storing yeast frozen at -80 degrees C in 15% glycerol. Glycerol stocks are good indefinitely (unless there are numerous freeze-thaws). When making a glycerol stock, we recommend using an overnight culture and concentrating it 2-4 fold. Spin down cells and suspend in 25-50% of the original volume with glycerol/medium. It is better to store frozen cells in fresh medium plus glycerol, rather than simply adding glycerol into the overnight culture.

    What choices do you offer for protein expression in a yeast host system, and what are their features?

    We offer the original Pichia pastoris expression systems, PichiaPink expression system, and Saccharomyces cerevisiae yeast expression system for expression of recombinant proteins. Both P. pastoris and S. cerevisiae have been genetically well-characterized and are known to perform many posttranslational modifications.

    The P. pastoris expression system combines the benefits of expression in E. coli (high-level expression, easy scale-up, and inexpensive growth) and the advantages of expression in a eukaryotic system (protein processing, folding, and posttranslational modifications), thus allowing high-level production of functionally active recombinant protein. As a yeast, Pichia pastoris shares the advantages of molecular and genetic manipulations with Saccharomyces cerevisiae, and it has the added advantage of 10- to 100-fold higher heterologous protein expression levels. These features make Pichia pastoris very useful as a protein expression system. The Pichia expression vectors contain either the powerful alcohol oxidase (AOX1) promoter for high-level, tightly controlled expression, or the glyceraldehyde-3-phosphate dehydrogenase (GAP) promoter for high-level, constitutive expression. Both inducible and constitutive expression constructs integrate into the P. pastoris genome, creating a stable host that generates extremely high protein expression levels, particularly when used in a fermentor. The Pichia pastoris expression systems we offer include:

    - PichiaPink Yeast Expression System: Newer Pichia pastoris expression system that contains both low- and high-copy plasmid backbones, 8 secretion signal sequences, and 4 yeast strains to help optimize for the highest yield possible of the recombinant protein. All PichiaPink vectors contain the AOX1 promoter for high-level, inducible expression and the ADE2 marker for selecting transformants using ADE2 complementation (i.e., by complementation of adenine auxotrophy) rather than antibiotic selection. However, they express the ADE2 gene product from promoters of different lengths, which dictate the copy number of the integrated plasmids. The pPink-LC vector has an 82 bp promoter for the ADE2marker and offers low-copy expression, and the pPink-HC vector has a 13 bp promoter for the ADE2marker and offers high-copy expression. The system also includes the pPinkalpha-HC vector (containing S. cerevisiae alpha-mating factor pre-sequence) for high copy number secreted expression, and provides eight secretion signal sequences for optimization of secreted expression.
    - EasySelect Pichia Expression Kit: One of the original Pichia expression kits that contains the pPICZ and pPICZalpha vectors, for intracellular and secreted expression, respectively, of the gene of interest. These vectors contain the AOX1 promoter for high-level, inducible expression and the Zeocin antibiotic resistance marker for direct selection of multi-copy integrants. They facilitate simple subcloning, simple purification, and rapid detection of expressed proteins.
    - Original Pichia Expression Kit: The kit includes the pPIC9, pPIC3.5, pHIL-D2, and pHIL-S1 vectors, each of which carries the AOX1 promoter for high-level, inducible expression and the HIS4 gene for selection in his4 strains, on histidine-deficient medium. pPIC9 carries the S. cerevisiae alpha-factor secretion signal while pHIL-S1 carries the Pichia pastoris alkaline phosphatase signal sequence (PHO) to direct transport of the protein to the medium. pHIL-D2 and pPIC3.5 are designed for intracellular expression.
    - Multi-Copy Pichia Expression Kit: This kit is designed to maximize expression and contains the pPIC3.5K, pPIC9K, and pAO815 vectors, which allow production and selection of Pichia strains that contain more than one copy of the gene of interest. They allow isolation and generation of multicopy inserts by in vivo methods (pPIC3.5K and pPIC9K) or in vitro methods (pAO815). All of these vectors contain the AOX1 promoter for high-level, inducible expression and the HIS4 gene for selection in his4 strains, on histidine-deficient medium. The pPIC9K vector directs secretion of expressed proteins while proteins expressed from pPIC3.5K and pAO815 remain intracellular. The pPIC9K and pPIC3.5K vectors carry the kanamycin resistance marker that confers resistance to Geneticin Reagent in Pichia. Spontaneous generation of multiple insertion events can be identified by resistance to increased levels of Geneticin Reagent. Pichia transformants are selected on histidine-deficient medium and screened for their level of resistance to Geneticin Reagent. The ability to grow in high concentrations of Geneticin indicates that multiple copies of the kanamycin resistance gene and the gene of interest are integrated into the genome.
    - For expression in S. cerevisiae, we offer the pYES Vector Collection. Each pYES vector carries the promoter and enhancer sequences from the GAL1 gene for inducible expression. The GAL1 promoter is one of the most widely used yeast promoters because of its strong transcriptional activity upon induction with galactose. pYES vectors also carry the 2m origin and are episomally maintained in high copy numbers (10-40 copies per cell).

    Why would I pick a yeast expression system for expression of my protein, as opposed to expression systems in other hosts?

    Yeast is a single-celled, eukaryotic organism that can grow quickly in defined media (doubling times are typically 2.5 hr in glucose-containing media) and is easier and less expensive to use for recombinant protein production than insect or mammalian cells (see table below). These positive attributes make yeast suitable for use in formats ranging from multi-well plates, shake flasks, and continuously stirred tank bioreactors to pilot plant and industrial-scale reactors.

    The most commonly employed species in the laboratory are Saccharomyces cerevisiae (also known as Baker's or Brewer's yeast) and some methylotrophic yeasts of the Pichia genus. Both S. cerevisiae and P. pastoris have been genetically characterized and shown to perform the posttranslational disulphide bond formation and glycosylation that is crucial for the proper functioning of some recombinant proteins. However, it is important to note that yeast glycosylation does differ from that in mammalian cells: in S. cerevisiae, O-linked oligosaccharides contain only mannose moieties, whereas higher eukaryotic proteins have sialylated O-linked chains. Furthermore S. cerevisiae is known to hyperglycosylate N-linked sites, which can result in altered protein binding, activity, and potentially yield an altered immunogenic response in therapeutic applications. In P. pastoris, oligosaccharides are of much shorter chain length and a strain has been reported that can produce complex, terminally sialylated or “humanized” glycoproteins.

    Do I need to include a ribosomal binding site (RBS/Shine Dalgarno sequence) or Kozak sequence when I clone my gene of interest?

    ATG is often sufficient for efficient translation initiation although it depends upon the gene of interest. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a Shine Dalgarno sequence/RBS or consensus Kozak sequence (ACCAUGG), as the case may be. In general, all expression vectors that have an N-terminal fusion will already have a RBS or initiation site for translation.

    Can the ProBond Purification system be used with Pichia lysates?

    Yes, you can use ProBond with His-tagged proteins expressed in Pichia. Here are some suggestions for using ProBond with Pichia supernatant:
    1. Adjust pH of Pichia supernatant to 7.5-8.0.
    2. Decant the supernatant from the heavy white precipitate. It is recommended to keep the precipitate for later solubilization in the rare case where the expressed protein has co-precipitated.
    3. Centrifuge the supernatant to remove leftover cell debris or other material that might clog the column.
    4. Adjust the conductivity to that of 500 mM NaCl with salt addition (may not be required since Pichia media is high salt).
    5. Run the column according to the instructions in the manual.

    Does Invitrogen have protocols or recommendations for Selenomethionine incorporation by Pichia to allow structural studies of crystallized protein?

    Jim Cregg (Keck Graduate Institute, developer of Pichia expression system for Phillips Petroleum), Senyon Choe (head of Structural Biology at The Salk Institute) and Thermo Fisher Scientific scientists have been unable to achieve adequate protein yields with greater than 40-50% incorporation of Selenomethionine (Se-Met), and unfortunately this level is not useful for phasing of crystals for x-ray analysis. The fundamental problem for Se-Met incorporation is that Met auxotrophic strains of Pichia, unlike Met auxotrophic strains of E. coli, do not grow on Se-Met. A number of groups have even attempted second site suppression mutagenesis in order to find genetic backgrounds that are Met auxotrophic but can grow (although poorly) on Se-Met. This has not been successful. Currently, Met WT strains of Pichia incorporate too much unmodified Met into proteins to produce protein that is >90% Se-Met substituted, which appears to be necessary for adequate phasing.

    Some promising references regarding this issue are:
    1. Larsson, et al. Preparation and crystallization of selenomethionyl dextranase from Penicillium minioluteum expressed in Pichia pastoris. Acta Crystallogr D Biol Crystallogr. 2002 Feb;58(Pt 2):346-8.
    2. Larsson, et al. Dextranase from Penicillium minioluteum: reaction course, crystal structure, and product complex. Structure (Camb). 2003 Sep;11(9):1111-21.
    3. Xu, et al. Crystallization and X-ray analysis of native and selenomethionyl beta-mannanase Man5A from blue mussel, Mytilus edulis, expressed in Pichia pastoris. Acta Crystallogr D Biol Crystallogr. 2002 Mar;58(Pt 3):542-5.

    How can I freeze Pichia?

    Store glycerol stocks frozen at -80° C in 15% glycerol. Glycerol stocks are good indefinitely (unless there are numerous freeze-thaws). When making a glycerol stock, we recommend using an overnight culture and concentrating it 2-4 fold. Spin down cells and suspend in 25-50% of the original volume with glycerol/medium. It is better to store frozen cells in fresh medium plus glycerol, rather than simply adding glycerol into the overnight culture.

    Are Invitrogen Pichia strains haploid or diploid? Can they sporulate?

    Pichia pastoris most commonly exists in a vegetative haploid state. On nitrogen limitation, mating can occur and diploid cells are formed. Since cells of the same strain can readily mate with each other, P. pastoris is by definition homothallic. Relative to Saccharomyces cerevisiae, which is heterothallic, the haploid state of P. pastoris is more stable. Under nitrogen limiting conditions P. pastoris diploids proceed through meiosis to the production of asci containing four haploid spores.

    Why are protease deficient Pichia pastoris and Pichia methanolica strains used for protein expression? What strains are available?

    DESCRIPTION OF PROTEOLYTIC ACTIVITIES
    Proteinase A is a vacuolar aspartyl protease capable of self-activation, as well as subsequent activation of additional vacuolar proteases, such as carboxypeptidase Y and proteinase B. Carobxypeptidase Y appears to be completely inactive prior to proteinase A-mediated proteolytic processing of the enzyme; proteinase B (encoded by the PrB gene of S. cerevisiae) reportedly is approximately 50% bioactive in its precursor form (i.e. the form that exists prior to proteinase A-mediated processing of the enzyme). Little is known about the proteolytic activities in Pichia pastoris. The following protease deficient Pichia pastoris strains have been made in an attempt to inactivate or delete the homologous proteolytic activities:

    SMD 1168 Pep4 gene disrupted
    SMD 1165 PrB gene disrupted
    SMD 1163 Pep4/PrB gene disrupted
    PichiaPink Strain 2 Pep4 gene disrupted
    PichiaPink Strain 3 Prb1 gene disrupted
    PichiaPink Strain 4 Prb1, Pep4 gene disrupted

    The Pep4 deficient mutant theoretically reduces the protease activity of Proteinase A, Caboxypeptisae Y, and approximately one-half of Proteinase B activity. The proteinase B deficient strain only reduces the activity of proteinase B. Finally, the Pep4/PrB strain reduces or eliminates the proteolytic activity of all three of these enzymes, proteinase A, Carboxypeptidase Y and Proteinase B. These protease deficient strains when compared to wild-type Pichia strains have shown to be highly efficient expression systems for the production of proteolytically sensitive products.

    The PRB1 deficient mutant is deficient in expression of proteinase B.

    PREPARATION OF PROTEASE DEFICIENT STRAINS
    The preferred method for preparing Pichia strains deficient in proteolytic activity, specific disruption of protease-encoding genes, was achieved by gene addition, gene replacement or a combination of additions and replacement referred to as "pop-in-pop-out" method. In gene replacement, the endogenous target gene is physically removed from the target locus, and replaced with a modified gene. This a accomplished by transforming the host with a linear fragment having ends which are homologous to the 5' and 3' ends of the target gene respectively. Gene addition involves adding the transforming DNA to the endogenous target gene. Depending on the manner in which the modified gene of the transforming DNA was altered, gene addition can result in the presence of either two non-functional copies of the target gene, or one functional and one non-functional copy of the target gene. Each of the two copies consists of a portion of the transforming DNA. If a functional copy of the target gene remains after gene addition, it can be removed by homologous recombination between the two copies of the target gene. The combination process of gene addition followed by homologous recombination constitutes the "Pop-in-pop-out" process.

    USEFULNESS
    When proteolytically sensitive recombinant products such as epidermal growth factor (EGF), growth hormone releasing factor (GRF), Insulin-like growth factor-1 (IGF-1), are expressed in Pichia strains which are deficient in proteolytic activity, higher levels of authentic bioactive recombinant product are produced. The following example illustrates the usefulness of these protease deficient strains:

    Proteolytic activity of the broth from a normal Pichia strain and a Pep4- Pichia strain were compared by adding known amount of peptide to the broth. The stability of the peptides in the two different broths was monitored by HPLC; when GRF or EFG were incubated with cell-free broth from these two strains, less than 10% of the peptides remained intact after 4 hours. In contrast, the GRF was greater than 60% intact after 4 hours and EGF remained greater than 90% after 8 hours incubation in the cell-free broth of the Pep4- strain. These data demonstrate that the disruption of the pep4 gene of Pichia result in a substantial reduction of the proteolyis.

    What is the mating genotype of Invitrogen Pichia strains, such as GS115 and SMD1168 and PichiaPink strain 1?

    All of our Pichia strains are homothallic strains. This means that they actually switch mating type with each generation. In Saccharomyces strains, this would lead to the culture rapidly becoming entirely diploid. In contrast, Pichia pastoris strains mate inefficiently to form diploids. Therefore, at any given time, the cells in the population are both a and alpha mating types.

    Do you need to add sulfuric acid to the Fermentation PTM Trace salts?

    You don't have to add sulfuric acid to your PTM1 salts or fermentation media. It would serve no purpose, other than may be help dissolve the salts.

    Can YPD be used instead of BMGY-type media for pichia fermentation?

    Yes. The cells will do fine in YPD but there are two drawbacks: The foaming that occurs in the richer YPD is very difficult to control. The richer media makes it difficult to purify secreted proteins from the media. The BMGY formulation remedies both of these problems.

    What can be used as an acid to adjust the pH of fermentation media? Does it even need to be pH'ed?

    No acid is required for Pichia fermentation. A healthy culture always acidifies the media. If the pH of the culture is increasing it is a sign of carbon source depletion or ill health of the culture.

    What pattern of oxygen uptake should I expect to observe during a fermentation run?

    It depends whether the clone is a Mut+ or a MutS.

    For a Mut+ you should expect that initially (in the first 2-4 hours of induction) the oxygen uptake rate of the culture would be lower than the end of glycerol batch phase. After the culture becomes adapted to methanol, the oxygen uptake rate will significantly increase, if the culture is healthy (i.e. not poisoned by too much methanol). One should run methanol spike tests during fermentation of Mut+ clones.

    For a MutS one can expect that the oxygen uptake rate will be lower than the end of glycerol batch phase through out most of the fermentation. One has to be very careful not to poison MutS clones.

    What is the Nitrogen source in the media for Pichia fermentation?

    During the Pichia fermentation process, the nitrogen source is the ammonium hydroxide used to adjust the pH. There is no nitrogen in the basal or trace salts.

    Can you tell me the difference between a Shine-Dalgarno sequence and a Kozak sequence?

    Prokaryotic mRNAs contain a Shine-Dalgarno sequence, also known as a ribosome binding site (RBS), which is composed of the polypurine sequence AGGAGG located just 5’ of the AUG initiation codon. This sequence allows the message to bind efficiently to the ribosome due to its complementarity with the 3’-end of the 16S rRNA. Similarly, eukaryotic (and specifically mammalian) mRNA also contains sequence information important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:

    - Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
    - Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
    - Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
    - Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
    - Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.

    Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:

    - Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
    - Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.


    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.