Learn More
CELLTREAT Scientific Products Pick n' ScoopTM Lab Spatula, Large, Non-sterile, Case Quantity 200
SDP
Catalog No. 5000670048
Encompass_Preferred
Dual-end lab spatula tool features a wide scoop for coarsemeasurements and narrow scoop with crafted point forprecise handling
Break up stubborn chemical clumps and transfer using oneconvenient tool
Multi-purpose bench tool saves space and enhances efficiency
Angled point is durable enough to break up tough, compacted,or hygroscopic chemicals without bending or breaking
Disposable spatulas reduce chemical cross contamination risks
U.S. patent pending
Catalog No. 50-006-70048 Supplier CELLTREAT Scientific Products Supplier No. 229970
Dual-end lab spatula tool features a wide scoop for coarsemeasurements and narrow scoop with crafted point forprecise handling
Break up stubborn chemical clumps and transfer using oneconvenient tool
Multi-purpose bench tool saves space and enhances efficiency
Angled point is durable enough to break up tough, compacted,or hygroscopic chemicals without bending or breaking
Disposable spatulas reduce chemical cross contamination risks
U.S. patent pending