Learn More
Vernier™ Picket Fence
SDPSCI_ED
Catalog No. S16058ND Shop All Vernier Software Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
S16058ND Plastic
1 options
Catalog No. S16058ND Supplier Vernier™ Supplier No. PF
Add to Cart
Add to Cart
Includes
Picket Fence
Picket Fences have eight opaque bars spaced every 5 cm, silk-screened directly onto clear plastic. Drop the picket fence through a photogate to obtain records of position, velocity and acceleration vs time or to measure g.
  • Eight opaque bars spaced every 5 cm, silk-screened directly onto clear plastic.
  • Drop the picket fence through a photogate to obtain records of position, velocity and acceleration vs time or to measure g.
Warranty and Services
5-year limited warranty
Specifications
Material Plastic
For Use With (Application) Educational use only
For Use With (Equipment) Go Direct™ Photogate, Vernier™ Photogate
Includes 8 opaque bars
Width (Metric) 5 cm bars