Learn More
Molecular Devices Molecular Devices QRep Replicator, 384 Pin, Polypropylene, 10 mm
Catalog No. 5000014581
Encompass_Preferred
The QRep 384-pin disposable replicator is a sterile, single-use polypropylene consumable designed for accurate transfer and replication of colonies and samples between microplates. It is intended for manual replication of clone libraries and for printing of microarrays in screening workflows.
  • Disposable polypropylene construction for chemical resistance and reduced cross-contamination.
  • 384 pins arranged in a 2 x 64 configuration for compatible 384-well plates.
  • 10 mm pin length suited for low-profile plate formats.
  • 1 mm pin diameter for precise transfer and minimal carryover.
  • Supplied sterile and packaged for single use.
  • Packaged in cases for laboratory-scale workflows.
Catalog No. 50-000-14581 Supplier Molecular Devices Supplier No. X5050
The QRep 384-pin disposable replicator is a sterile, single-use polypropylene consumable designed for accurate transfer and replication of colonies and samples between microplates. It is intended for manual replication of clone libraries and for printing of microarrays in screening workflows.
  • Disposable polypropylene construction for chemical resistance and reduced cross-contamination.
  • 384 pins arranged in a 2 x 64 configuration for compatible 384-well plates.
  • 10 mm pin length suited for low-profile plate formats.
  • 1 mm pin diameter for precise transfer and minimal carryover.
  • Supplied sterile and packaged for single use.
  • Packaged in cases for laboratory-scale workflows.