Learn More
Molecular Devices Molecular Devices Automated Cell Imaging System, Brightfield, Fluorescence, Digital Confocal
Catalog No. 5000014671
Encompass_Preferred
Compact automated cell imaging and analysis system for fast acquisition and entry-level high-content workflows. Supports brightfield, fluorescence, and Digital Confocal 2D imaging with autofocus and live preview, and uses icon-driven software with preconfigured protocols to simplify common cell-based assays.
  • Automated brightfield, fluorescence, and Digital Confocal 2D imaging.
  • User-exchangeable objectives with a 6-position automated turret.
  • Standard 4x NA 0.13 and 10x NA 0.32 objectives; optional 20x, 40x, and 63x available.
  • User-exchangeable filter cubes; standard FITC included; additional DAPI, CFP, TRITC, and Cy5 options.
  • Icon-driven acquisition and analysis software with preconfigured protocols for common assays.
  • Supports 6- to 384-well microplates, slides, and 35 mm culture dishes.
  • Optional environmental control for live-cell, time-lapse experiments.
Catalog No. 50-000-14671 Supplier Molecular Devices Supplier No. PICOBASIC
Add to Cart
Add to Cart
Request a Quote
Compact automated cell imaging and analysis system for fast acquisition and entry-level high-content workflows. Supports brightfield, fluorescence, and Digital Confocal 2D imaging with autofocus and live preview, and uses icon-driven software with preconfigured protocols to simplify common cell-based assays.
  • Automated brightfield, fluorescence, and Digital Confocal 2D imaging.
  • User-exchangeable objectives with a 6-position automated turret.
  • Standard 4x NA 0.13 and 10x NA 0.32 objectives; optional 20x, 40x, and 63x available.
  • User-exchangeable filter cubes; standard FITC included; additional DAPI, CFP, TRITC, and Cy5 options.
  • Icon-driven acquisition and analysis software with preconfigured protocols for common assays.
  • Supports 6- to 384-well microplates, slides, and 35 mm culture dishes.
  • Optional environmental control for live-cell, time-lapse experiments.