Learn More
Research Products International Corp Pico Methyl-Seq Library Prep Kit, 10 Preps
SDP
Catalog No. 501977024
Encompass
The Pico Methyl-Seq Library Prep Kit provides a streamlined workflow for making WGBS libraries. Input DNA is randomly fragmented during the initial bisulfite treatment step followed by three rounds of amplification with uniquely designed primers. The procedure can accommodate as little as 10 pg input DNA (including that derived from FFPE samples), making it ideal for methylation analysis of precious, limited, and target-enriched samples.
All-inclusive kit for bisulfite conversion followed by Whole Genome Bisulfite Sequencing (WGBS) library preparation.
Accommodates ultra-low DNA input and compatible with FFPE samples.
Simple, ligation- and gel-free workflow can be completed in a few hours.
Catalog No. 50-197-7024 Supplier Research Products International Corp Supplier No. ZD5455
The Pico Methyl-Seq Library Prep Kit provides a streamlined workflow for making WGBS libraries. Input DNA is randomly fragmented during the initial bisulfite treatment step followed by three rounds of amplification with uniquely designed primers. The procedure can accommodate as little as 10 pg input DNA (including that derived from FFPE samples), making it ideal for methylation analysis of precious, limited, and target-enriched samples.
All-inclusive kit for bisulfite conversion followed by Whole Genome Bisulfite Sequencing (WGBS) library preparation.
Accommodates ultra-low DNA input and compatible with FFPE samples.
Simple, ligation- and gel-free workflow can be completed in a few hours.