Learn More
Zymo Research Corporation Pico Methyl-Seq™ Library Prep Kit (10 preps.) -[Includes Room Temp Item & Dry Ice Item] '**'
SDP
Catalog No. NC0680296
Encompass_Preferred
The Pico Methyl-Seq Library Prep Kit provides a streamlined workflow for making WGBS libraries. Input DNA is randomly fragmented during the initial bisulfite treatment step followed by three rounds of amplification with uniquely designed primers. The procedure can accommodate as little as 10 pg input DNA (including that derived from FFPE samples), making it ideal for methylation analysis of precious, limited, and target-enriched samples.
Catalog No. NC0680296 Supplier Zymo Research Corporation Supplier No. D5455
The Pico Methyl-Seq Library Prep Kit provides a streamlined workflow for making WGBS libraries. Input DNA is randomly fragmented during the initial bisulfite treatment step followed by three rounds of amplification with uniquely designed primers. The procedure can accommodate as little as 10 pg input DNA (including that derived from FFPE samples), making it ideal for methylation analysis of precious, limited, and target-enriched samples.