Learn More
Lumiprobe Pico488 dsDNA quantification reagent, 200x solution in DMSO, 100 uL
Catalog No. NC1878927
Encompass_Preferred
Pico488 is a DNA binding benzothiazole dye whose structure is identical to PicoGreen. This reagent is a 200× solution in DMSO. The dye is a fluorescent sensor that is specific for double stranded DNA (dsDNA). Its absorption and emission in the fluorescein channel can be easily detected using cuvette fluorometers, well plate readers, and by dedicated DNA measurement fluorometers like Qubit.
Catalog No. NC1878927 Supplier Lumiprobe Supplier No. 12010
Pico488 is a DNA binding benzothiazole dye whose structure is identical to PicoGreen. This reagent is a 200× solution in DMSO. The dye is a fluorescent sensor that is specific for double stranded DNA (dsDNA). Its absorption and emission in the fluorescein channel can be easily detected using cuvette fluorometers, well plate readers, and by dedicated DNA measurement fluorometers like Qubit.